2-Fluoro-4-(trifluoromethoxy)benzoic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
H33749.03EA
112
EUR
H33749.03
H33749.06
2-Fluoro-4-(trifluoromethoxy)benzoic acid ≥97%
2-Fluoro-4-(trifluoromethoxy)benzoic acid
Formula:
C₈H₄F₄O₃ MW: 224,11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16652439 CAS Number: 1073477-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...