3-Fluoro-4-(trifluoromethoxy)benzoic acid ≥97%

Supplier: Thermo Scientific
Warning

H26234.06EA 372 EUR
H26234.06 H26234.03
3-Fluoro-4-(trifluoromethoxy)benzoic acid ≥97%
3-Fluoro-4-(trifluoromethoxy)benzoic acid
Formula: C₈H₄F₄O₃
Melting Pt: 98…101 °C
Storage Temperature: Ambient
MDL Number: MFCD06660202
CAS Number: 886498-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR