4-Fluoro-2-(trifluoromethoxy)benzoic acid ≥97%
|
Warning
|
H33363.03EA
98.6
EUR
H33363.03
H33363.06
4-Fluoro-2-(trifluoromethoxy)benzoic acid ≥97%
4-Fluoro-2-(trifluoromethoxy)benzoic acid
Formula:
C₈H₄F₄O₃ MW: 224,11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687173 CAS Number: 1242250-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...