4-Fluoro-3-(trifluoromethoxy)benzoic acid ≥97%

Supplier: Thermo Scientific

Warning

H26368.06EA 321 EUR
H26368.06 H26368.03
4-Fluoro-3-(trifluoromethoxy)benzoic acid ≥97%
4-Fluoro-3-(trifluoromethoxy)benzoic acid
Formula: C₈H₄F₄O₃
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD06660161
CAS Number: 886496-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR