Benzyl chloromethyl ether

Supplier: Molekula

MOLEM21886700EA 0 EUR
MOLEM21886700
Benzyl chloromethyl ether
Benzyl chloromethyl ether
Formula: C₈H₉ClO
MW: 156,61
MDL Number: MFCD00000886
CAS Number: 3587-60-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR