2-(Bromomethyl)benzothiazole

MAYBCC06008DAEA 0 EUR
MAYBCC06008DA MAYBCC06008CB
2-(Bromomethyl)benzothiazole
2-(Bromomethyl)benzothiazole
Formula: C₈H₆BrNS
MW: 228,11
MDL Number: MFCD00461195
CAS Number: 106086-78-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR