4-Benzyl-4-hydroxypiperidine 95%

Warning

ACRO105990050EA 88.8 EUR
ACRO105990050
4-Benzyl-4-hydroxypiperidine 95%
4-Benzyl-4-hydroxypiperidine
Formula: C₁₂H₁₇NO
Melting Pt: 80…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00023147
CAS Number: 51135-96-7
EINECS: 257-003-7

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
GC ≥94 %
Water ≤0.05 %
GC ≥99.0 %
Color scale ≤50 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR