3-(3-(Trifluoromethoxy)phenyl)propionic acid 95%

Warning

B21233.01EA 49 EUR
B21233.01 B21233.03 B21233.06
3-(3-(Trifluoromethoxy)phenyl)propionic acid 95%
3-(3-(Trifluoromethoxy)phenyl)propionic acid
Formula: C₁₀H₉F₃O₃
MW: 234,17
Storage Temperature: Ambient
MDL Number: MFCD01317834
CAS Number: 168833-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR