3-(4-(Difluoromethoxy)phenyl)propionic acid 97%, JRD

Supplier: Thermo Scientific

Warning

H32381.MDEA 58.8 EUR
H32381.MD H32381.03
3-(4-(Difluoromethoxy)phenyl)propionic acid 97%, JRD
3-(4-(Difluoromethoxy)phenyl)propionic acid
Formula: C₁₀H₁₀F₂O₃
MW: 216,18
Melting Pt: 66…69 °C
Storage Temperature: Ambient
MDL Number: MFCD09863355
CAS Number: 1000932-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR