2-Amino-N-isopropylbenzamide, Supelco®
36941-250MGEA
0
EUR
36941-250MG
2-Amino-N-isopropylbenzamide, Supelco®
2-Amino-N-isopropylbenzamide
Formula:
C₁₀H₁₄N₂O |
MDL Number:
MFCD00143620 CAS Number: 30391-89-0 EINECS: 250-175-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...