2-Chloro-5-(pentafluorothio)benzaldehyde 97%

Supplier: Thermo Scientific

Warning

H35247.03EA 155 EUR
H35247.03 H35247.MD
2-Chloro-5-(pentafluorothio)benzaldehyde 97%
2-Chloro-5-(pentafluorothio)benzaldehyde
Formula: C₇H₄ClF₅OS
MW: 266,61
Storage Temperature: Ambient
MDL Number: MFCD22988924
CAS Number: 1431329-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR