2-Methyl-5-(trifluoromethoxy)anisole ≥97%
|
Warning
|
H33477.03EA
155
EUR
H33477.03
H33477.MD
2-Methyl-5-(trifluoromethoxy)anisole ≥97%
2-Methyl-5-(trifluoromethoxy)anisole
Formula:
C₉H₉F₃O₂ MW: 206,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18399698 CAS Number: 1261444-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...