4'-Methylacetanilide

Synonyms: p-AcetotoluididN-(4-Methylphenyl)acetamidep-Acetotoluidide

MAYBRH01049.1EA 0 EUR
MAYBRH01049.1 CHMPRH01049.250
4'-Methylacetanilide
4'-Methylacetanilide
Formula: C₉H₁₁NO
MW: 149,19 g/mol
Boiling Pt: 307 °C (1013 hPa)
Melting Pt: 145…148 °C
Density: 1,22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008677
CAS Number: 103-89-9
EINECS: 203-155-4
Merck Index: 13,00076

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR