(+)-Isomenthol

Supplier: Merck

59802-25MGEA 146 EUR
59802-25MG
(+)-Isomenthol
(+)-Isomenthol
Formula: C₁₀H₂₀O
CAS Number: 23283-97-8
EINECS: 245-554-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR