1,3-Benzoxazol-6-amine 95%

Supplier: Thermo Scientific

Danger

Synonyms: 6-Aminobenzoxazole

H34485.MDEA 81 EUR
H34485.MD H34485.03 H34485.06
1,3-Benzoxazol-6-amine 95%
1,3-Benzoxazol-6-amine
Formula: C₇H₆N₂O
MW: 134,10
Melting Pt: 95…97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03791187
CAS Number: 177492-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR