Zero air generators, ZA range, Peak Scientific
564-0107EA 0 EUR
PEAK30-1020 PEAK30-1560 PEAK30-1670 PEAK30-1610 PEAK30-1060 PEAK30-1170 PEAK30-1570 PEAK30-1620 PEAK30-1230 PEAK30-1160 PEAK30-1110 PEAK30-1010 PEAK30-1660 PEAK30-1120
Zero air generators, ZA range, Peak Scientific
The ZA range of zero air generators is designed specifically for use as flame gas for GC applications which require zero air for operation. The catalyst chambers inside these zero air generators make sure that compressed air is stripped of all hydrocarbons before it is fed into your application.
  • Forced air ventilation, self-checks for leaks and automatic shut-down function
  • Units are available with or without internal air compressors
  • Total hydrocarbon concentration <0,1 ppm