Earmuffs, Leightning® L2
Supplier: Howard Leight by Honeywell
Leightning®
111-1406EA 28.6 EUR
111-1406
Earmuffs, Leightning® L2
Earmuffs Noise Blocking Earmuffs
Robust and durable earmuffs for rough industrial environments.
  • Medium profile cups
  • Air Flow Control™ Technology provides better, consistent attenuation, especially at low frequencies
  • Snap in ear cushions makes cushion replacement quick and easy
  • Padded foam headband delivers long wearing comfort and reduces the pressure on the head
  • Telescopic height adjustment remains fixed during use
The earmuffs work especially well in extreme temperatures/humidity and under aggressive chemical influences.
SNR: 31 dB (headband)
SNR: 32 dB (foldable)
Weight: 231 g (headband); 236 g (foldable)
Certifications: EN 352-1