Dry block thermostat for cuvettes, DB-10C

Supplier: Grant

GRANDB-10CEA 397 EUR
GRANDB-10C
Dry block thermostat for cuvettes, DB-10C
Dry block thermostat which holds 10 cuvettes (each with 10 mm optical pathway), maintains constant temperature of the samples to be measured. The integral aluminium block provides excellent stability and uniformity. The easy to use unit has a 2-line LCD display which shows both set and actual values for temperature and time. The DB-10C is ideal for preparation of photometric analysis and diagnostic methods (enzyme reaction intensity and metabolite concentration).

  • Operating temperature range is ambient + 5 to 42 °C
  • Dry temperature control system maintains clean and aerosol free environment
  • Temperature stability and uniformity at 37 °C are ±0,1 and ±0,3 °C
  • Timer with audible alarm, 1 to 96 h

Delivery information: Supplied with block for 10 cuvettes.

* RT = Ambient
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR