2-Amino-4-(4-bromophenyl)-5-methylthiazole 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-(4-Bromophenyl)-5-methylthiazol-2-amine
H58657.06EA
271
EUR
H58657.06
2-Amino-4-(4-bromophenyl)-5-methylthiazole 97%
2-Amino-4-(4-bromophenyl)-5-methylthiazole
Formula:
C₁₀H₉BrN₂S MW: 269,16 g/mol Melting Pt: 151…153 °C |
MDL Number:
MFCD00170268 CAS Number: 65705-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...