Methylmagnesium chloride 3 M in THF

Supplier: Thermo Scientific

Danger


Packaged under argon in resealable ChemSeal™ bottles.

42101.JSEA 38.6 EUR
42101.JS 42101.JU
Methylmagnesium chloride 3 M in THF
Methylmagnesium chloride
Formula: CH₃ClMg
MW: 74,79 g/mol
Density: 1,013
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000468
CAS Number: 676-58-4
EINECS: 211-629-7
UN: 3399
ADR: 4.3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR