Methylmagnesium chloride 3 M in THF

Danger

46118.JUEA 155 EUR
46118.JU
Methylmagnesium chloride 3 M in THF
Methylmagnesium chloride

As a grignard reagent and an alkylating agent in organic synthesis

Formula: CH₃ClMg
MW: 74,79 g/mol
Density: 1,013
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000468
CAS Number: 676-58-4
EINECS: 211-629-7
UN: 3399
ADR: 4.3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR