Ebselen

Supplier: Enzo Life Sciences
Danger

Synonyms: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one2-Phenylbenzo[d][1,2]selenazol-3(2H)-one

ALEXALX270097M025EA 148 EUR
ALEXALX270097M025 ALEXALX270097M005
Ebselen
Ebselen
Formula: C₁₃H₉NOSe
MW: 274,20 g/mol
MDL Number: MFCD00210937
CAS Number: 60940-34-3
UN: 3283
ADR: 6.1,III
https://ie.vwr.com/stibo/web/std.lang.all/74/75/45347475.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR