1,4-Dioxene ≥98%
Supplier: Thermo Scientific
|
Danger
|
H33597.06EA
0
EUR
H33597.06
1,4-Dioxene ≥98%
1,4-Dioxene
Formula:
C₄H₆O₂ MW: 86,09 g/mol Boiling Pt: 94…96 °C Density: 1,08 Flash Pt: 1 °C (34 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00082537 CAS Number: 543-75-9 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...