Tetramethytetraselenafulvalene 97%

Supplier: Thermo Scientific

Danger

Synonyms: Tetramethyltetraselenafulvalene

ACRO346500500EA 74 EUR
ACRO346500500 ACRO346502500
Tetramethytetraselenafulvalene 97%
Tetramethytetraselenafulvalene
Formula: C₁₀H₁₂Se₂
Melting Pt: 265…270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011507
CAS Number: 55259-49-9
UN: 3283
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Red to violet Crystalline powder
NMR (1H-NMR) Consistent with structure
NMR (1H-NMR) ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR