3-Cyano-6-methylchromone ≥99%

Danger

Synonyms: 6-Methylchromone-3-carbonitrile

H27825.03EA 35.3 EUR
H27825.03 H27825.06
3-Cyano-6-methylchromone ≥99%
3-Cyano-6-methylchromone
Formula: C₁₁H₇NO₂
MW: 185,19
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00191962
CAS Number: 50743-18-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR