4'-(1-Imidazolyl)acetophenone ≥97%

Supplier: Thermo Scientific

L06115.06EA 54.7 EUR
L06115.06 L06115.14
4'-(1-Imidazolyl)acetophenone ≥97%
4'-(1-Imidazolyl)acetophenone
Formula: C₁₁H₁₀N₂O
MW: 186,21
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00005282
CAS Number: 10041-06-2
EINECS: 233-123-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR