1-(Phenylsulphonyl)-3-indoleboronic acid
Supplier: Molekula
Synonyms:
(1-(Phenylsulphonyl)-1H-indol-3-yl)boronic acid
MOLE59160967-1GEA
0
EUR
MOLE59160967-1G
1-(Phenylsulphonyl)-3-indoleboronic acid
1-(Phenylsulphonyl)-3-indoleboronic acid
Formula:
C₁₄H₁₂BNO₄S MW: 301,1300 g/mol |
MDL Number:
MFCD02681892 CAS Number: 129271-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...