4-(2-Carboxyvinyl)phenylboronic acid 95%

Supplier: Alfa Aesar
Warning

Synonyms: 4-(2-Carboxyvinyl)benzeneboronic acid

L15586.06EA 463 EUR
L15586.06 L15586.03 L15586.MD
4-(2-Carboxyvinyl)phenylboronic acid 95%
4-(2-Carboxyvinyl)phenylboronic acid
Formula: C₉H₉BO₄
Melting Pt: ∼ 190…196 °C
Storage Temperature: Ambient
MDL Number: MFCD01075704
CAS Number: 151169-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR