N,N'-Diallyl-L-tartardiamide

Supplier: Molekula

MOLE14688583-100GEA 0 EUR
MOLE14688583-100G
N,N'-Diallyl-L-tartardiamide
N,N'-Diallyl-L-tartardiamide
Formula: C₁₀H₁₆N₂O₄
MW: 228,25
MDL Number: MFCD00008640
CAS Number: 58477-85-3
EINECS: 261-277-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR