Dodecyl methyl sulphide

Supplier: Thermo Scientific

14877.06EA 40.9 EUR
14877.06
Dodecyl methyl sulphide
Dodecyl methyl sulphide
Formula: C₁₃H₂₈S
MW: 216,41
Boiling Pt: 163…165 °C (19 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00015073
CAS Number: 3698-89-3
EINECS: 223-028-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR