4-(2,5-Dimethylphenylcarbamoyl)benzeneboronic acid, 96%

H52850.MDEA 27.1 EUR
ALFAH52850.MD ALFAH52850.03
4-(2,5-Dimethylphenylcarbamoyl)benzeneboronic acid, 96%
Purity: 96%
CAS No.: 913835-40-2
Molecular Formula: C15H16BNO3
Formula Weight: 269.11
MDL No.: MFCD09027214
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR