2-Chloro-4-(2,3-dimethylphenylcarbamoyl)benzeneboronic acid, 97%

H54696.03EA 206.4 EUR
ALFAH54696.03 ALFAH54696.MD
2-Chloro-4-(2,3-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR