2-Chloro-5-(ethoxycarbonyl)benzeneboronic acid, 98%

H53031.MDEA 31.9 EUR
ALFAH53031.MD ALFAH53031.03
2-Chloro-5-(ethoxycarbonyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 913835-93-5
Molecular Formula: C9H10BClO4
Formula Weight: 228.44
Melting Point: 236-240°
MDL No.: MFCD08689529
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR