2-Chloro-4-(4-fluoro-2-nitrophenylcarbamoyl)benzeneboronic acid, 97%

H55554.MDEA 29.4 EUR
ALFAH55554.MD ALFAH55554.03
2-Chloro-4-(4-fluoro-2-nitrophenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C13H9BClFN2O5
Formula Weight: 338.48
MDL No.: MFCD20040314
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR