5-Dimethylcarbamoyl-3-fluorobenzeneboronic acid, 98%

H53100.MDEA 52.3 EUR
ALFAH53100.MD ALFAH53100.03
5-Dimethylcarbamoyl-3-fluorobenzeneboronic acid, 98%
Purity: 98%
CAS No.: 874219-39-3
Molecular Formula: C9H11BFNO3
Formula Weight: 211.00
Melting Point: 111-113°
MDL No.: MFCD08235056
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR