4-(2,3-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

H55782.MDEA 60.3 EUR
ALFAH55782.MD ALFAH55782.03
4-(2,3-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040326
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR