3-(2,4-Dimethylphenylcarbamoyl)-4-fluorobenzeneboronic acid, 97%

H54453.03EA 147.4 EUR
ALFAH54453.03 ALFAH54453.MD
3-(2,4-Dimethylphenylcarbamoyl)-4-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040189
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR