3-Fluoro-5-(2-methylphenylcarbamoyl)benzeneboronic acid, 97%

H54183.03EA 289.7 EUR
ALFAH54183.03 ALFAH54183.MD
3-Fluoro-5-(2-methylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H13BFNO3
Formula Weight: 273.07
MDL No.: MFCD20265200
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR