5-Fluoro-(2-methoxycarbonyl)phenylboronic acid ≥97%

Supplier: Thermo Scientific

Warning

Synonyms: 5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid

H52756.MDEA 0 EUR
H52756.MD
5-Fluoro-(2-methoxycarbonyl)phenylboronic acid ≥97%
5-Fluoro-(2-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BFO₄
MW: 197,96
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD03095367
CAS Number: 850568-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR