5-(Ethoxycarbonyl)-2-fluorophenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Ethoxycarbonyl-2-fluorobenzeneboronic acid

H53134.MDEA 76.2 EUR
H53134.MD H53134.03
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid ≥97%
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid
Formula: C₉H₁₀BFO₄
MW: 211,98
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD08235094
CAS Number: 874219-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR