3-Chloro-5-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%

Supplier: Thermo Scientific

Synonyms: 3-Chloro-5-(trifluoromethyl)benzeneboronic acid pinacol ester

H51712.03EA 232 EUR
H51712.03 H51712.MD
3-Chloro-5-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%
3-Chloro-5-(trifluoromethyl)phenylboronic acid pinacol ester
Formula: C₁₃H₁₅BClF₃O₂
MW: 306,52
Storage Temperature: Ambient
MDL Number: MFCD11856039
CAS Number: 942069-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR