3'-Chloro-5'-(trifluoromethoxy)acetophenone ≥97%

Supplier: Thermo Scientific

Warning

H26503.03EA 97.5 EUR
H26503.03 H26503.06
3'-Chloro-5'-(trifluoromethoxy)acetophenone ≥97%
3'-Chloro-5'-(trifluoromethoxy)acetophenone
Formula: C₉H₆ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06660315
CAS Number: 886503-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR