2-Chloro-6-phenoxybenzylamine
MAYBCD11381CBEA
0
EUR
MAYBCD11381CB
MAYBCD11381DA
2-Chloro-6-phenoxybenzylamine
2-Chloro-6-phenoxybenzylamine
Formula:
C₁₃H₁₂ClNO |
MDL Number:
MFCD00052915 CAS Number: 175136-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...