4-Chlorobenzyl-2-hydroxyethyl sulphide ≥98%

Supplier: Thermo Scientific

Warning

Synonyms: 2-(4-Chlorobenzylthio)ethanol

L10158.06EA 74.4 EUR
L10158.06 L10158.14
4-Chlorobenzyl-2-hydroxyethyl sulphide ≥98%
4-Chlorobenzyl 2-hydroxyethyl sulphide
Formula: C₉H₁₁ClOS
Boiling Pt: 139…140 °C (1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00020614
CAS Number: 71501-40-1
EINECS: 275-568-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR