2'-Hydroxy-5'-(trifluoromethoxy)acetophenone ≥98%

Supplier: Thermo Scientific
Warning

H32003.03EA 136 EUR
H32003.03 H32003.06 H32003.MD
2'-Hydroxy-5'-(trifluoromethoxy)acetophenone ≥98%
2'-Hydroxy-5'-(trifluoromethoxy)acetophenone
Formula: C₉H₇F₃O₃
Melting Pt: 28…31 °C
Storage Temperature: Ambient
MDL Number: MFCD01091002
CAS Number: 146575-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR