4-Methoxy-2-(trifluoromethyl)benzamide ≥97%

Supplier: Thermo Scientific
Warning

H32608.06EA 304 EUR
H32608.06 H32608.03
4-Methoxy-2-(trifluoromethyl)benzamide ≥97%
4-Methoxy-2-(trifluoromethyl)benzamide
Formula: C₉H₈F₃NO₂
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD09025404
CAS Number: 916420-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR