1,3-Diphenylacetone-p-toluenesulphonylhydrazone ≥99%

Supplier: Thermo Scientific

L03625.18EA 180 EUR
L03625.18 L03625.09
1,3-Diphenylacetone-p-toluenesulphonylhydrazone ≥99%
1,3-Diphenylacetone p-toluenesulphonylhydrazone
Formula: C₂₂H₂₂N₂O₂S
Melting Pt: ∼ 185 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00009645
CAS Number: 19816-88-7
EINECS: 243-344-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR