3-Benzylcarbamoyl-4-chlorobenzeneboronic acid, 97%

H54263.MDEA 60.3 EUR
ALFAH54263.MD ALFAH54263.03
3-Benzylcarbamoyl-4-chlorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H13BClNO3
Formula Weight: 289.52
MDL No.: MFCD20265217
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR