2-Chloro-5-(4-chlorobenzylcarbamoyl)benzeneboronic acid, 97%

H54528.03EA 144.9 EUR
ALFAH54528.03 ALFAH54528.MD
2-Chloro-5-(4-chlorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BCl2NO3
Formula Weight: 323.97
MDL No.: MFCD27975652
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR