2-Fluoro-4-biphenylboronic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Fluoro-1,1'-biphenyl-4-ylboronic acid
, (3-fluoro-4-phenyl)phenylboronic acid
, 3-Fluoro-4-phenylbenzeneboronic acid
, 2-Fluoro-4-biphenylylboronic acid
L15634.06EA
142
EUR
L15634.06
2-Fluoro-4-biphenylboronic acid ≥97%
2-Fluoro-4-biphenylboronic acid
Formula:
C₁₂H₁₀BFO₂ MW: 216,02 g/mol Melting Pt: ∼ 242…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075707 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...